missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABL4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100569
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total IFT27.
RABL4 belongs to the small GTPase superfamily, Ras family. RABL4 possesses GTPase activity. This gene encodes a putative GTP-binding protein similar to RAY/RAB1C. The protein is ras-related, but the function is unknown.
Specifications
| RABL4 | |
| Polyclonal | |
| Unconjugated | |
| IFT27 | |
| 2600013G09Rik; BBS19; IFT27; intraflagellar transport 27; intraflagellar transport 27 homolog; intraflagellar transport protein 27 homolog; Putative GTP-binding protein RAY-like; RAB, member of RAS oncogene family-like 4; RABL4; rab-like protein 4; RAS oncogene family-like 4; RAYL | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11020, 67042 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BW83, Q9D0P8 | |
| IFT27 | |
| A synthesized peptide derived from human IFT27(Accession Q9BW83), corresponding to amino acid residues K47-F97. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction