missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557178
This item is not returnable.
View return policy
Description
Immunogen sequence: LAEDFNPEEI NLDCTNPRYL AAGSSNAVKL SRFFDKVIEK RYFLREGNQI PGFPDRKRFG AGTLKSLHYD Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
Predicted to enable GTP binding activity; GTPase activity; and protein homodimerization activity. Involved in regulation of Ras protein signal transduction and regulation of protein lipidation. Predicted to be active in endomembrane system. Implicated in pancreatic cancer.
Specifications
| RABL3 | |
| Polyclonal | |
| Unconjugated | |
| RABL3 | |
| 4930553C05Rik; AA958834; RAB, member of RAS oncogene family like 3; RAB, member of RAS oncogene family-like 3; RAB, member RAS oncogene family-like 3; Rabl3; rab-like protein 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 285282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5HYI8 | |
| RABL3 | |
| Recombinant protein corresponding to Human RABL3. Recombinant protein control fragment (Product #RP-96363). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction