missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RABIF Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584226
This item is not returnable.
View return policy
Description
Immunogen sequence: GDLLQEHWLV EDMFIFENVG FTKDVGNIKF LVCADCEIGP IGWHCLDDKN SFYVALERVS HE Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 97%.
The Sec4/Rab-related small GTP-binding proteins are involved in the regulation of intracellular vesicular transport. Mss4 stimulates GTP-GDP exchange in Sec4 and Rab and binds to a subset of genetically related Rab proteins.
Specifications
| RABIF | |
| Polyclonal | |
| Unconjugated | |
| RABIF | |
| AI842864; AW549708; E130318E12Rik; Guanine nucleotide exchange factor MSS4; guanine-nucleotide-releasing protein mss4; mammalian suppressor of SEC4; Mss4; RAB interacting factor; RABIF; RAB-interacting factor; RASGFR3; RASGRF3; Ras-specific guanine-releasing factor 3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P47224 | |
| RABIF | |
| Recombinant protein corresponding to Human RABIF. Recombinant protein control fragment (Product #RP-108406). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction