missing translation for 'onlineSavingsMsg'
Learn More

RABIF/MSS4 Antibody (2G3), Novus Biologicals™

Product Code. 18368868 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368868 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18368868 Supplier Novus Biologicals Supplier No. H00005877M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RABIF/MSS4 Monoclonal antibody specifically detects RABIF/MSS4 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABIF/MSS4
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2G3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH18488
Gene Alias mammalian suppressor of SEC4, mss4, MSS4guanine nucleotide exchange factor MSS4, RAB interacting factor, Rab-interacting factor, RASGFR3, RASGRF3, Ras-specific guanine-releasing factor 3
Host Species Mouse
Immunogen RABIF (AAH18488, 1 a.a. ~ 123 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEPAEQPSELVSAEGRNRKAVLCQRCGSRVLQPGTALFSRRQLFLPSMRKKPALSDGSNPDGDLLQEHWLVEDMFIFENVGFTKDVGNIKFLVCADCEIGPIGWHCLDDKNSFYVALERVSHE
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5877
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.