missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabies Virus Nucleoprotein Antibody (12) - (CVS-11), Novus Biologicals™
Mouse Monoclonal Antibody
£145.00 - £295.00
Specifications
| Antigen | Rabies Virus Nucleoprotein |
|---|---|
| Clone | 12 |
| Dilution | ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA, Sandwich ELISA Detection |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30160540
|
Novus Biologicals (Bio-Techne)
NBP3-33166-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30170633
|
Novus Biologicals (Bio-Techne)
NBP3-33166-100ul |
100 μL |
£295.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rabies Virus Nucleoprotein Monoclonal antibody specifically detects Rabies Virus Nucleoprotein in Virus samples. It is validated for ELISA,Sandwich ELISA DetectionSpecifications
| Rabies Virus Nucleoprotein | |
| ELISA 1:1000-1:2000, Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| IgG2a | |
| Protein A purified |
| 12 | |
| ELISA, Sandwich ELISA Detection | |
| Unconjugated | |
| Mouse | |
| Virus | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rabies Virus Nucleoprotein (Uniprot#: Q8JXF6; Met1-Ser450). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title