missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabies Virus Monoclonal Antibody (1C5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA121549
This item is not returnable.
View return policy
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Rabies virus (Neurotropic lyssavirus) is a member of the Rhabdoviridae family. Rabies is a single stranded, neurotropic, negative sense RNA virus which encodes 5 proteins: a glycoprotein, a nucleoprotein, and three others. The mature virus has a bullet shape, a protein coat, and a lipid envelope. The outer surface of the virus is covered with thumblike glycoprotein projections 5-10 nm long and 3 nm in diameter. The virus averages approximately 780 nm in length. Virus infectivity is destroyed by lipid solvents. Rabies virus is a very successful virus, with a very wide range of hosts. It causes an acute, central nervous system infection, characterized by CNS irritation, followed by paralysis and death. Approximately 50,000 human deaths each year are caused by rabies.
Specifications
| Rabies Virus | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2a |
| ELISA, Immunohistochemistry | |
| 1C5 | |
| Unconjugated | |
| Neurotropic lyssavirus | |
| Russian rabies vaccine strain Vnukovo-32. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction