missing translation for 'onlineSavingsMsg'
Learn More

RABGGTB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£145.00 - £417.00

Specifications

Antigen RABGGTB
Dilution Western Blot 1:200 - 1:2000, ELISA
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30227135
View Documents
Novus Biologicals
NBP3-35528-100ul
100 μL
£417.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30226593
View Documents
Novus Biologicals
NBP3-35528-20ul
20 μL
£145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

RABGGTB Polyclonal antibody specifically detects RABGGTB in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications

Specifications

RABGGTB
ELISA, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
EC 2.5.1.60, Geranylgeranyl transferase type II subunit beta, geranylgeranyl transferase type-2 subunit beta, GGTase-II-beta, GGTB, Rab geranylgeranyltransferase subunit beta, Rab geranyl-geranyltransferase subunit beta, Rab geranylgeranyltransferase, beta subunit, Rab GG transferase beta, Rab GGTase beta, Type II protein geranyl-geranyltransferase subunit beta
Recombinant fusion protein containing a sequence corresponding to amino acids 1-331 of human RABGGTB (NP_004573.2).,, Sequence:, MGTPQKDVIIKSDAPDTLLLEKHADYIASYGSKKDDYEYCMSEYLRMSGIYWGLTVMDLMGQLHRMNREEILAFIKSCQHECGGISASIGHDPHLLYTLSAVQILTLYDSINVIDVNKVVEYVKGLQKEDGSFAGDIWGEIDTRFSFCAVATLALLGKLDAINVEKAIEFVLSCMNFDGGFGCRPGSESHAGQIYCCTGFLAITSQLHQVNSDLLGWWLCERQLPSGGLNGRPEKLPDVCYSWWVLASLKIIGRLHWIDREKLRNFILACQDEETGGFADRPGDMVDPFHTLFGIAGLSLLGEEQIKPVNPVFCMPEEVLQRVNVQPELVS
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:200 - 1:2000, ELISA
Polyclonal
Purified
RUO
PBS (pH 7.3), 50% glycerol
5876
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.