missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGGTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14448-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the transfer of a geranylgeranyl moiety from geranylgeranyl diphosphate to both cysteines of Rab proteins with the C-terminal sequence -XXCC, -XCXC and -CCXX, such as RAB1A, RAB3A, RAB5A and RAB7A.Specifications
| RABGGTA | |
| Polyclonal | |
| Unconjugated | |
| RABGGTA | |
| PTAR3, Rab GG transferase alpha, Rab GGTase alpha, RABGGTA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 56187, 5875, 58983 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| 0.45 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q08602, Q92696, Q9JHK4 | |
| Rabggta | |
| RABGGTA Fusion Protein Ag5923 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction