missing translation for 'onlineSavingsMsg'
Learn More

RABGGTA Antibody, Novus Biologicals™

Product Code. 18349068 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349068 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349068 Supplier Novus Biologicals Supplier No. H00005875B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

RABGGTA Polyclonal antibody specifically detects RABGGTA in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABGGTA
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_004572
Gene Alias EC 2.5.1.60, Geranylgeranyl transferase type II subunit alpha, geranylgeranyl transferase type-2 subunit alpha, protein prenyltransferase alpha subunit repeat containing 3, PTAR3, Rab geranylgeranyltransferase subunit alpha, Rab geranyl-geranyltransferase subunit alpha, Rab geranylgeranyltransferase, alpha subunit, Rab GG transferase alpha, Rab GGTase alpha
Host Species Mouse
Immunogen RABGGTA (NP_004572, 1 a.a. - 567 a.a.) full-length human protein. MHGRLKVKTSEEQAEAKRLEREQKLKLYQSATQAVFQKRQAGELDESVLELTSQILGANPDFATLWNCRREVLQQLETQKSPEELAALVKAELGFLESCLRVNPKSYGTWHHRCWLLGRLPEPNWTRELELCARFLEVDERNFHCWDYRRFVATQAAVPPAEELAFTDSLITRNFSNYSSWHYRSCLLPQLHPQPDSGPQGRLPEDVLLKELELVQNAFFTDPNDQSAWFYHRWLLGRADPQDALRCLHVSRDEACLTVSFSRPLLVGSRMEILLLMVDDSPLIVEWRTPDGRNRPSHVWLCDLPAASLNDQLPQHTFRVIWTAGDVQKECVLLKGRQEGWCRDSTTDEQLFRCELSVEKSTVLQSELESCKELQELEPENKWCLLTIILLMRALDPLLYEKETLQYFQTLKAVDPMRATYLDDLRSKFLLENSVLKMEYAEVRVLHLAHKDLTVLCHLEQLLLVTHLDLSHNRLRTLPPALAALRCLEVLQASDNAIESLDGVTNLPRLQELLLCNNRLQQPAVLQPLASCPRLVLLNLQGNPLCQAVGILEQLAELLPSVSSVLT
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5875
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.