missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABGAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35604-100ul
This item is not returnable.
View return policy
Description
RABGAP1 Polyclonal antibody specifically detects RABGAP1 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| RABGAP1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp586D2123, GAP and centrosome-associated protein, GAPCENA, RAB GTPase activating protein 1, rab GTPase-activating protein 1, rab6 GTPase activating protein (GAP and centrosome-associated), Rab6 GTPase activating protein, GAPCenA, Rab6 GTPase-activating protein GAPCenA, RP11-123N4.2, TBC1 domain family, member 11, TBC1D11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 830-1069 of human RABGAP1 (NP_036329.3).,, Sequence:, QQEDPIERFERENRRLQEANMRLEQENDDLAHELVTSKIALRKDLDNAEEKADALNKELLMTKQKLIDAEEEKRRLEEESAQLKEMCRRELDKAESEIKKNSSIIGDYKQICSQLSERLEKQQTANKVEIEKIRQKVDDCERCREFFNKEGRVKGISSTKEVLDEDTDEEKETLKNQLREMELELAQTKLQLVEAECKIQDLEHHLGLALNEVQAAKKTWFNRTLSSIKTATGVQGKETC | |
| 100 μL | |
| Cell Cycle and Replication | |
| 23637 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction