missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabex-5 Polyclonal Antibody
Description
A suggested positive control is rat brain tissue lysate. PA5-21117 can be used with blocking peptide PEP-1231.
RABGEF1 forms a complex with rabaptin-5 (RABPT5) that is required for endocytic membrane fusion, and it serves as a specific guanine nucleotide exchange factor (GEF) for RAB5.
Specifications
Specifications
| Antigen | Rabex-5 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | RABGEF1 |
| Gene Accession No. | Q9JM13, Q9UJ41 |
| Gene Alias | BTB/POZ domain-containing protein KCTD7; RAB guanine nucleotide exchange factor (GEF) 1; RAB guanine nucleotide exchange factor 1; Rab5 exchange factor; rab5 GDP/GTP exchange factor; Rab5 GDP/GTP exchange factor, Rabex5; Rab5 guanine nucleotide exchange factor Rabex-5; Rab5ef; Rabaptin-5-associated exchange factor for Rab5; Rabex5; rabex-5; RABEX5 protein; RABGEF1; RAP1; Ras negative regulator Rabex-5; Ras negative regulator Rabex-5/Rin2; Rin2; zgc:65944 |
| Gene Symbols | RABGEF1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?