missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole, TRC

Product Code. 30135897
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135897 500 mg 500mg
30011745 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135897 Supplier Toronto Research Chemicals Supplier No. TRCR070540500MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117976-89-3
Molecular Formula C18 H21 N3 O3 S
Molecular Weight (g/mol) 359.44
Synonym 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-, 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole, (±)-Rabeprazole, 2-[[[3-Methyl-4-(3-methoxypropoxy)-2-pyridinyl]methyl]sulfinyl]-1H-benzimidazole, 2-[[[3-Methyl-4-(3-methoxypropoxy)-2-pyridyl]methyl]sulfinyl]-1H-benzimidazole, 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methyl]sulfinyl]-1H-benzimidazole, 2-[[[4-(3-Methoxypropoxy)-3-methyl-2-pyridyl]methyl]sulfinyl]benzimidazole, 2-[[[4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methyl]sulfinyl]-1H-benzimidazole, Dexrabeprazol, Habeprazole, LY 307640, Newprazole, Pariets, Rabeloc, Rabemax, Rabeprazole, Rabiral
IUPAC Name 2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMILES COCCCOc1ccnc(CS(=O)c2nc3ccccc3[nH]2)c1C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C18H21N3O3S/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,20,21)
Formula Weight 359.1304
Chemical Name or Material Rabeprazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.