missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sulfone, TRC

Product Code. 30010280
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010280 25 mg 25mg
30003883 50 mg 50mg
17901615 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010280 Supplier Toronto Research Chemicals Supplier No. TRCR07050525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117976-47-3
Molecular Formula C18 H21 N3 O4 S
Molecular Weight (g/mol) 375.44
Synonym Rabeprazole Sulphone
IUPAC Name 2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfonyl]-1H-benzimidazole
SMILES COCCCOc1ccnc(CS(=O)(=O)c2nc3ccccc3[nH]2)c1C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H21N3O4S/c1-13-16(19-9-8-17(13)25-11-5-10-24-2)12-26(22,23)18-20-14-6-3-4-7-15(14)21-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,20,21)
Formula Weight 375.1253
Chemical Name or Material Rabeprazole Sulfone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.