missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sulfone N-Oxide, TRC

Product Code. 30412096
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30412096 10 mg
30413109 25 mg
30419293 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30412096 Supplier Toronto Research Chemicals Supplier No. TRCR07051510MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 924663-37-6
Molecular Formula C18 H21 N3 O5 S
Molecular Weight (g/mol) 391.44
Synonym Rabeprazole Sulphone N-Oxide, 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-1-oxido-2-pyridinyl]methyl]sulfonyl]-, 2-[[[4-(3-Methoxypropoxy)-3-methyl-1-oxido-2-pyridinyl]methyl]sulfonyl]-1H-benzimidazole, Rabeprazole sulfone N-oxide
IUPAC Name 2-[[4-(3-methoxypropoxy)-3-methyl-1-oxidopyridin-1-ium-2-yl]methylsulfonyl]-1H-benzimidazole
SMILES COCCCOc1cc[n+]([O-])c(CS(=O)(=O)c2nc3ccccc3[nH]2)c1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H21N3O5S/c1-13-16(21(22)9-8-17(13)26-11-5-10-25-2)12-27(23,24)18-19-14-6-3-4-7-15(14)20-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,19,20)
Formula Weight 391.1202
Percent Purity >95%
Chemical Name or Material Rabeprazole Sulfone N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.