missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole Sodium Salt, TRC

Product Code. 17924235
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17924235 25 mg 25mg
17952615 50 mg 50mg
17961835 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17924235 Supplier Toronto Research Chemicals Supplier No. TRCR07050025MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117976-90-6
Molecular Formula C18 H20 N3 O3 S . Na
Molecular Weight (g/mol) 381.42
Synonym 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-, sodium salt (1:1), 1H-Benzimidazole, 2-[[[4-(3-methoxypropoxy)-3-methyl-2-pyridinyl]methyl]sulfinyl]-, sodium salt (9CI), 2-[[[4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methyl]sulfinyl]-1H-benzimidazole sodium salt, Aciphex, E 3810, LY 307640 sodium, Pariet, Pariprazole, Pepcia, Rabeprazole sodium, Rabicip, Razo
IUPAC Name sodium;2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl]benzimidazol-1-ide
SMILES [Na+].COCCCOc1ccnc(CS(=O)c2nc3ccccc3[n-]2)c1C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1
Formula Weight 381.1123
Percent Purity >95%
Chemical Name or Material Rabeprazole Sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.