missing translation for 'onlineSavingsMsg'
Learn More

Rabeprazole sodium, MedChemExpress

Product Code. p-200096392
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16464437

Brand: Medchem Express HYB0656A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Rabeprazole sodium (LY307640 sodium) is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. Rabeprazole sodium induces apoptosis. Rabeprazole sodium acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. Rabeprazole sodium can be used for the research of gastric ulcerations and gastroesophageal reflux.

Rabeprazole sodium (LY307640 sodium) is a second-generation proton pump inhibitor (PPI) that irreversibly inactivates gastric H+/K+-ATPase. Rabeprazole sodium induces apoptosis. Rabeprazole sodium acts as an uridine nucleoside ribohydrolase (UNH) inhibitor with an IC50 of 0.3 μM. Rabeprazole sodium can be used for the research of gastric ulcerations and gastroesophageal reflux.

Specifications

Chemical Name or Material Rabeprazole sodium
CAS 117976-90-6
Color White
Purity Grade Notes Research
Molecular Formula C18H20N3NaO3S
Quantity 50 mg
Synonym LY307640 sodium
Solubility Information H2O : ≥ 100 mg/mL (262.18 mM) ∣DMSO : ≥ 48 mg/mL (125.85 mM)
SMILES O=S(C1=NC2=CC=CC=C2N1[Na])CC3=NC=CC(OCCCOC)=C3C
Molecular Weight (g/mol) 381.42
Formula Weight 381.42
Percent Purity 99.09%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.