missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabeprazole EP Impurity A (Rabeprazole USP Related Compound D), Mikromol™
Description
Rabeprazole EP Impurity A (Rabeprazole USP Related Compound D)
Specifications
Specifications
| Chemical Name or Material | Rabeprazole Sulfone |
| CAS | 117976-47-3 |
| Analyte or Component Names | Rabeprazole Sulfone |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H21 N3 O4 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Rabeprazole Sulphone, Rabeprazole Sodium Imp. A (EP), Rabeprazole Imp. A (EP), Rabeprazole USP Related Compound D, Rabeprazole USP RC D:, Rabeprazole Related Compound D, Rabeprazole Sodium Impurity A, Rabeprazole Impurity A |
| InChI Formula | InChI=1S/C18H21N3O4S/c1-13-16(19-9-8-17(13)25-11-5-10-24-2)12-26(22,23)18-20-14-6-3-4-7-15(14)21-18/h3-4,6-9H,5,10-12H2,1-2H3,(H,20,21) |
| SMILES | COCCCOc1ccnc(CS(=O)(=O)c2nc3ccccc3[nH]2)c1C |
| IUPAC Name | 2-[[4-(3-methoxypropoxy)-3-methylpyridin-2-yl]methylsulfonyl]-1H-benzimidazole |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?