missing translation for 'onlineSavingsMsg'
Learn More

RABEP1 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18401117 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401117 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401117 Supplier Novus Biologicals Supplier No. NBP147285G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RABEP1 Polyclonal antibody specifically detects RABEP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen RABEP1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias neurocrescin, rab GTPase-binding effector protein 1, RAB5EPRABPT5A, rabaptin, RAB GTPase binding effector protein 1, rabaptin-4, rabaptin-5, Rabaptin-5alpha, RABPT5rabaptin-5alpha, Renal carcinoma antigen NY-REN-17
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human Rabaptin, RAB GTPase Binding Effector Protein 1 using the numbering given in entry NP_004694.2 (GeneID 9135).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9135
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.