missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rabenosyn 5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15778671 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15778671 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15778671 Supplier Invitrogen™ Supplier No. PA560853

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DSPAPNPFSE EDEHPQQRLS SPLVPGNPFE EPTCINPFEI DSDSGPEAEE PIEEELLLQQ IDNIKAYIFD AKQC Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 80%.

This gene encodes a zinc finger transcription factor. The encoded protein likely plays a role in transcriptional repression of interleukin 2. Mutations in this gene have been associated with posterior polymorphous corneal dystrophy-3 and late-onset Fuchs endothelial corneal dystrophy. Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabenosyn 5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Rbsn
Gene Accession No. Q9H1K0
Gene Alias 110 kDa protein; 5330426D11Rik; FLJ34993; FYVE finger-containing Rab5 effector protein rabenosyn-5; MGC126210; RAB effector RBSN; Rabenosyn 5; rabenosyn, RAB effector; rabenosyn-5; Rbsn; ZFYVE20; zinc finger FYVE domain-containing protein 20; zinc finger, FYVE domain containing 20
Gene Symbols Rbsn
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Rabenosyn 5. Recombinant protein control fragment (Product #RP-96334).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64145
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.