missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabenosyn 5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-33649
This item is not returnable.
View return policy
Description
Rabenosyn 5 Polyclonal specifically detects Rabenosyn 5 in Human samples. It is validated for Western Blot.
Specifications
| Rabenosyn 5 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| RBSN | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Rabenosyn 5. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 64145 | |
| Human | |
| IgG |
| Western Blot | |
| 0.98mg/mL | |
| Western Blot 1:500-1:3000 | |
| 110 kDa protein, FLJ34993, FYVE finger-containing Rab5 effector protein rabenosyn-5, FYVE-finger-containing Rab5 effector protein rabenosyn-5, MGC126210, Rabenosyn-5, Zinc finger FYVE domain-containing protein 20, zinc finger, FYVE domain containing 20 | |
| Rabbit | |
| 89 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction