Learn More
Rabbit IgG Isotype Control (RbNP15), eBioscience™, Invitrogen™
Rabbit Recombinant Monoclonal Isotype Control
Brand: Invitrogen 14-4616-82
Additional Details : Weight : 0.10000kg
Description
Description: This product is a rabbit monoclonal IgG and can be used as a matched isotype control for recombinant rabbit monoclonal antibodies. The RbNP15 IgG was selected as an isotype control following screening for low background on a variety of mouse and human cell types. Applications Tested: This rabbit monoclonal isotype control has been tested for use as a negative control for flow cytometric analysis. Please use at the same concentration as the experimental antibody. Filtration: 0.2 μm post-manufacturing filtered.
Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.Specifications
Rabbit IgG | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Rabbit | |
100 μg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Control, Flow Cytometry | |
RbNP15 | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG; Not Applicable | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |