missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit IgG Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems BAF008
This item is not returnable.
View return policy
Description
IgG Polyclonal specifically detects IgG in Rabbit samples. It is validated for Western Blot, Simple Western.
Specifications
| IgG | |
| Polyclonal | |
| Biotin | |
| Immunoglobulin G, ImmunoglobulinG | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3500 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot | |
| IGHG1 | |
| Rabbit IgG | |
| 250 μg | |
| Secondary | |
| Detects rabbit IgG heavy and light chains. In direct ELISAs, less than 5% cross-reactivity with human IgG, mouse IgG and chicken IgY is observed. | |
| Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction