Learn More
ZNF407, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZNF407.
Brand: Abnova PAB22124.100uL
Description
This gene encodes a zinc finger protein whose exact function is not known. It may be involved in transcriptional regulation. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: PVLGNKEILMNSQHETEFILEEDGPASDSTVESSDVYETIISIDDKGQAMYSFGRFDSSIIRIKNPEDGELIDQSEEGLIATGVRISELPLKDCAQGVKKKKSEGSSIGESTRIRCDDCGFLSpecifications
| ZNF407 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZNF407. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ13839/FLJ20307/KIAA1703 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55628 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| ZNF407 | |
| ZNF407 | |
| Recombinant protein corresponding to amino acids of human ZNF407. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only