missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF320, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZNF320.
Brand: Abnova PAB24052.100uL
This item is not returnable.
View return policy
Description
ZNF320 encodes a Kruppel-like zinc finger protein. Members of this protein family are involved in activation or repression of transcription.[supplied by OMIM
Sequence: GKVFSLRSLLAEHQKIPFGDNCFKCNEYSKPSSISpecifications
| ZNF320 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZNF320. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp686G16228/ZFPL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 162967 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| ZNF320 | |
| ZNF320 | |
| Recombinant protein corresponding to amino acids of human ZNF320. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction