missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZMYM2 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZMYM2.
Brand: Abnova PAB19403.100ug
This item is not returnable.
View return policy
Specifications
| ZMYM2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (0. 125-0.25 ug/mL) Immunocytochemistry (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| ZMYM2 | |
| ZMYM2 | |
| A synthetic peptide corresponding to 17 amino acids near C-terminus of human ZMYM2. | |
| 100 μg | |
| Primary | |
| At least two isoforms of ZMYM2 are known to exist; this antibody will detect only the shorter isoform. This antibody is predicted to not cross-react with other ZMYM protein family members. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunocytochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of ZMYM2. | |
| In PBS (0.02% sodium azide) | |
| FIM/MYM/RAMP/SCLL/ZNF198 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 7750 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction