Learn More
ZMIZ2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZMIZ2.
Brand: Abnova PAB23684.100uL
Description
ZMIZ2 and ZMIZ1 (MIM 607159) are members of a PIAS (see MIM 603566)-like family of proteins that interact with nuclear hormone receptors. ZMIZ2 interacts with androgen receptor (AR; MIM 313700) and enhances AR-mediated transcription (Huang et al., 2005 [PubMed 16051670]).[supplied by OMIM
Sequence: LMPSVMEMIAALGPGAAPFAPLQPPSVPAPSDYPGQGSSFLGPGTFPESFPPTTPSTPTLAEFTPGPPPISYQSDIPSSLLTSEKSTACLPSQMAPAGHLDPTHNPGTPGLHTSNLSpecifications
| ZMIZ2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZMIZ2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp761I2123/KIAA1886/ZIMP7/hZIMP7 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 83637 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ZMIZ2 | |
| ZMIZ2 | |
| Recombinant protein corresponding to amino acids of human ZMIZ2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only