Learn More
ZIC3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZIC3.
Brand: Abnova PAB1954.400uL
Description
This gene encodes a member of the ZIC family of C2H2-type zinc finger proteins. This nuclear protein probably functions as a transcription factor in early stages of left-right body axis formation. Mutations in this gene cause X-linked visceral heterotaxy, which includes congenital heart disease and left-right axis defects in organs. [provided by RefSeq
Specifications
| ZIC3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ZIC3. | |
| In PBS (0.09% sodium azide) | |
| HTX/HTX1/ZNF203 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 7547 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| ZIC3 | |
| ZIC3 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ZIC3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only