missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zhx2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant Zhx2.
Brand: Abnova PAB15774.50ug
This item is not returnable.
View return policy
Specifications
| Zhx2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant Zhx2. | |
| In PBS (50% glycerol, 0.02% sodium azide) | |
| Afr-1/Afr1/Raf/mKIAA0854 | |
| Rabbit | |
| 50 μg | |
| Primary | |
| Specific to recombinant protein GX0354. This antibody detects mZHX2 protein. It also recognizes human ZHX2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Zhx2 | |
| Zhx2 | |
| Recombinant GST fusion protein corresponding to 121 amino acids of mouse Zhx2. | |
| RUO | |
| 387609 | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction