Learn More
ZFY, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZFY.
Brand: Abnova PAB20081.100uL
Description
This gene encodes a zinc finger-containing protein that may function as a transcription factor. This gene was once a candidate gene for the testis-determining factor (TDF) and was erroneously referred to as TDF. [provided by RefSeq
Sequence: IGPDGHPLTVYPCMICGKKFKSRGFLKRHMKNHPEHLAKKKYHCTDCDYTTNKKISLHNHLESHKLTSKAEKAIECDECGKHFSHAGALFTHKMVHKEKGANKMHKCKFCEYETAEQGLLNRHLSpecifications
| ZFY | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZFY. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| MGC138710 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7544 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| ZFY | |
| ZFY | |
| Recombinant protein corresponding to amino acids of human ZFY. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only