Learn More
ZDHHC15, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZDHHC15.
Brand: Abnova PAB21089.100uL
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardatio X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: VSRNKTTLEAFCTPVFTSGPEKNGFNLGFIKNIQQVFGDKKKFWLIPIGSSPGDGHSFPMRSMNESQNPLLANEETWEDNEDDNQSpecifications
| ZDHHC15 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZDHHC15. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ31812/MGC119974/MGC119975/MGC119976/MRX91 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 158866 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| ZDHHC15 | |
| ZDHHC15 | |
| Recombinant protein corresponding to amino acids of human ZDHHC15. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only