missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBED5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant ZBED5.
Brand: Abnova PAB18531.100ug
This item is not returnable.
View return policy
Specifications
| ZBED5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant ZBED5. | |
| In buffer containing 0.02% sodium azide | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 58386 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user. | |
| BC047754 | |
| Recombinant protein corresponding to amino acids 224-474 of human ZBED5. | |
| 100 μg | |
| Primary | |
| This antibody is specific to ZBED5. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction