Learn More
YWHAZ Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of YWHAZ.
Brand: Abnova PAB4028.400uL
Description
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and sheep orthologs. The encoded protein interacts with IRS1 protein, suggesting a role in regulating insulin sensitivity. Several transcript variants that differ in the 5' UTR but that encode the same protein have been identified for this gene. [provided by RefSeq]
Specifications
| YWHAZ | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of YWHAZ. | |
| In PBS (0.09% sodium azide) | |
| KCIP-1/MGC111427/MGC126532/MGC138156 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 7534 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:50-100) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| YWHAZ | |
| YWHAZ | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding T232 of human YWHAZ. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only