Learn More
YWHAQ Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of YWHAQ.
Brand: Abnova PAB14303.200ug
Description
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse and rat orthologs. This gene is upregulated in patients with amyotrophic lateral sclerosis. It contains in its 5' UTR a 6bp tandem repeat sequence which is polymorphic, however, there is no correlation between the repeat number and the disease. [provided by RefSeq]
Specifications
| YWHAQ | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of YWHAQ. | |
| In PBS (0.2% gelatin, 0.09% sodium azide) | |
| 14-3-3/1C5/HS1 | |
| Rabbit | |
| 200 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:400) The optimal working dilution should be determined by the end user. | |
| YWHAQ | |
| YWHAQ | |
| A synthetic peptide corresponding to human YWHAQ. | |
| RUO | |
| 10971 | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only