Learn More
XYLT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant XYLT1.
Brand: Abnova PAB20403.100uL
Description
Xylosyltransferase (XT; EC 2.4.2.26) catalyzes the transfer of UDP-xylose to serine residues within XT recognition sequences of target proteins. Addition of this xylose to the core protein is required for the biosynthesis of the glycosaminoglycan chains characteristic of proteoglycans.[supplied by OMIM
Sequence: LITLETQDGYFSHRPKEKVRTDSNNENSVPKDFENVDNSNFAPRTQKQKHQPELAKKPPSRQKELLKRKLEQQEKGKGHTFPGKGPGEVLPPGDRAAANSSHGKDVSRPSpecifications
| XYLT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant XYLT1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| XT-I/XT1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 64131 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| XYLT1 | |
| XYLT1 | |
| Recombinant protein corresponding to amino acids of human XYLT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only