Learn More
XRCC6 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of XRCC6.
Brand: Abnova PAB27138.100uL
Description
The p70/p80 autoantigen is a nuclear complex consisting of two subunits with molecular masses of approximately 70 and 80kDa. The complex functions as a single-stranded DNA-dependent ATP-dependent helicase. The complex may be involved in the repair of nonhomologous DNA ends such as that required for double-strand break repair, transposition, and V(D)J recombination. High levels of autoantibodies to p70 and p80 have been found in some patients with systemic lupus erythematosus. [provided by RefSeq
Specifications
| XRCC6 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| XRCC6 | |
| XRCC6 | |
| A synthetic peptide corresponding to human XRCC6. | |
| 100 μL | |
| Primary | |
| XRCC6 polyclonal antibody detects endogenous levels of XRCC6 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of XRCC6. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CTC75/CTCBF/G22P1/KU70/ML8/TLAA | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 2547 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only