Learn More
XRCC3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of XRCC3.
Brand: Abnova PAB24835.100uL
Description
This gene encodes a member of the RecA/Rad51-related protein family that participates in homologous recombination to maintain chromosome stability and repair DNA damage. This gene functionally complements Chinese hamster irs1SF, a repair-deficient mutant that exhibits hypersensitivity to a number of different DNA-damaging agents and is chromosomally unstable. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. Alternatively spliced transcript variants encoding the same protein have been identified. [provided by RefSeq
Specifications
| XRCC3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| XRCC3 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 7517 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of XRCC3. | |
| In 1X PBS, pH 7.2 (0.05% sodium azide) | |
| XRCC3 | |
| A synthetic peptide corresponding to XRCC3. | |
| 100 μL | |
| Primary | |
| XRCC3 polyclonal antibody detects endogenous levels of XRCC3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only