Learn More
XAF1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of XAF1.
Brand: Abnova PAB13125.100ug
Description
X-linked inhibitor of apoptosis (XIAP; MIM 300079) is a potent member of the IAP family. All members of this family possess baculoviral IAP (BIR) repeats, cysteine-rich domains of approximately 80 amino acids that bind and inhibit caspases (e.g., CASP3; MIM 600636). XIAP has 3 BIR domains and a C-terminal RING zinc finger that possesses E3 ubiquitin ligase (see MIM 601623) activity. XAF1 antagonizes the anticaspase activity of XIAP and may be important in mediating apoptosis resistance in cancer cells (Liston et al., 2001 [PubMed 11175744]).[supplied by OMIM
Specifications
| XAF1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of XAF1. | |
| In PBS (0.02% sodium azide) | |
| BIRC4BP/HSXIAPAF1/XIAPAF1 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Multiple isoforms of XAF-1 exist and a doublet at ∼30 KDa can be seen using anti-XAF-1 (IN). | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| XAF1 | |
| XAF1 | |
| A synthetic peptide corresponding to internal region 16 amino acids of human XAF1. | |
| RUO | |
| 54739 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only