Learn More
WNT7A Rabbit anti-Bats, Bovine, Dog, Hamster, Horse, Human, Monkey, Mouse, Pig, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyconal antibody raised against synthetic peptide of WNT7A.
Brand: Abnova PAB28369.50ug
Description
This gene is a member of the WNT gene family, which consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is involved in the development of the anterior-posterior axis in the female reproductive tract, and also plays a critical role in uterine smooth muscle pattering and maintenance of adult uterine function. Mutations in this gene are associated with Fuhrmann and Al-Awadi/Raas-Rothschild/Schinzel phocomelia syndromes. [provided by RefSeq
Specifications
| WNT7A | |
| Polyclonal | |
| Rabbit polyconal antibody raised against synthetic peptide of WNT7A. | |
| In PBS (0.09% sodium azide) | |
| WNT7A | |
| A synthetic peptide corresponding to 10 amino acids at internal region of human WNT7A. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/ml) The optimal working dilution should be determined by the end user. | |
| WNT7A | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 7476 | |
| Bat, Bovine, Canine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only