Learn More
WNT5B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WNT5B.
Brand: Abnova PAB16482.50ug
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 94% and 80% amino acid identity to the mouse Wnt5b protein and the human WNT5A protein, respectively. Alternative splicing of this gene generates 2 transcript variants. [provided by RefSeq
Specifications
| WNT5B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of WNT5B. | |
| In PBS (0.09% sodium azide) | |
| MGC2648 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 81029 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2.5 ug/mL) The optimal working dilution should be determined by the end user. | |
| WNT5B | |
| WNT5B | |
| A synthetic peptide (conjugated with KLH) corresponding to human WNT5B. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only