Learn More
WNT2B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WNT2B.
Brand: Abnova PAB16477.50ug
Description
This gene encodes a member of the wingless-type MMTV integration site (WNT) family of highly conserved, secreted signaling factors. WNT family members function in a variety of developmental processes including regulation of cell growth and differentiation and are characterized by a WNT-core domain. This gene may play a role in human development as well as human carcinogenesis. This gene produces two alternatively spliced transcript variants. [provided by RefSeq
Specifications
| WNT2B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of WNT2B. | |
| In PBS (0.09% sodium azide) | |
| WNT13/XWNT2 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 7482 | |
| Bovine, Canine, Equine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| WNT2B | |
| WNT2B | |
| A synthetic peptide (conjugated with KLH) corresponding to human WNT2B. | |
| 50 μg | |
| Primary | |
| Internal domain of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only