Learn More
WNT2 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WNT2.
Brand: Abnova PAB26053.50ug
Description
This gene is a member of the WNT gene family. The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. Alternatively spliced transcript variants have been identified for this gene. [provided by RefSeq
Specifications
| WNT2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of WNT2. | |
| In PBS (0.09% sodium azide) | |
| INT1L1/IRP | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 7472 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| WNT2 | |
| WNT2 | |
| A synthetic peptide corresponding to 15 amino acids at internal region of human WNT2. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only