Learn More
WNT16 Rabbit anti-Hamster, Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WNT16.
Brand: Abnova PAB26056.50ug
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It contains two transcript variants diverging at the 5' termini. These two variants are proposed to be the products of separate promoters and not to be splice variants from a single promoter. They are differentially expressed in normal tissues, one of which (variant 2) is expressed at significant levels only in the pancreas, whereas another one (variant 1) is expressed more ubiquitously with highest levels in adult kidney, placenta, brain, heart, and spleen. [provided by RefSeq]
Specifications
| WNT16 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of WNT16. | |
| In PBS (0.09% sodium azide) | |
| WNT16 | |
| A synthetic peptide corresponding to 16 amino acids at internal region of human WNT16. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins, except CETN3 (50%). | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
| WNT16 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 51384 | |
| Hamster, Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only