Learn More
WIPF2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WIPF2.
Brand: Abnova PAB21701.100uL
Description
This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery. [provided by RefSeq
Sequence: PPPYRMHGSEPPSRGKPPPPPSRTPAGPPPPPPPPLRNGHRDSITTVRSFLDDFESKYSFHPVEDFPAPEEYKHFQSpecifications
| WIPF2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WIPF2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| WICH/WIRE | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 147179 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| WIPF2 | |
| WIPF2 | |
| Recombinant protein corresponding to amino acids of human WIPF2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only