missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR48, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WDR48.
Brand: Abnova PAB23285.100uL
This item is not returnable.
View return policy
Description
Sequence: SIIQCHILNDKRHILTKDTNNNVAYWDVLKACKVEDLGKVDFEDEIKKRFKMVYVPNWFSVDLKTGMLTITLDESDCFAAWVSAKDAGFSpecifications
| WDR48 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WDR48. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp686G1794/KIAA1449/P80 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 57599 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| WDR48 | |
| WDR48 | |
| Recombinant protein corresponding to amino acids of human WDR48. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction