Learn More
WDR31 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WDR31.
Brand: Abnova PAB21202.100uL
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene but the biological validity of some variants has not been determined. [provided by RefSeq
Sequence: REPYILQTSEDKTLRLWDSRGLQVAHMFPAKQHIQTYCEVSVDGHKCISCSNGFGGEGCEATLWDLRQTRNRICEYKGHFQTVASpecifications
| WDR31 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WDR31. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ35921 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 114987 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| WDR31 | |
| WDR31 | |
| Recombinant protein corresponding to amino acids of human WDR31. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only