Learn More
WDR3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WDR3.
Brand: Abnova PAB22044.100uL
Description
This gene encodes a nuclear protein containing 10 WD repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, which usually include a trp-asp at the C-terminal end. Proteins belonging to the WD repeat family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. [provided by RefSeq
Sequence: AKEDQPAVPGETQGDSYFTGKKTIETVKAAERIMEAIELYREETAKMKEHKAICKAAGKEVPLPSNPILMAYGSISPSAYVLEIFKGIKSSELEESLLVLPSpecifications
| WDR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WDR3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| FLJ12796 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 10885 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| WDR3 | |
| WDR3 | |
| Recombinant protein corresponding to amino acids of human WDR3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only