Learn More
WDR26, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WDR26.
Brand: Abnova PAB22739.100uL
Description
This gene encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. Two transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Sequence: NDLNELKPLVHSPHAIVRMKFLLLQQKYLEYLEDGKVLEALQVLRCELTPLKYNTERIHVLSGYLMCSHAEDLRAKAEWESpecifications
| WDR26 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant WDR26. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CDW2/FLJ21016/MIP2 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 80232 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| WDR26 | |
| WDR26 | |
| Recombinant protein corresponding to amino acids of human WDR26. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only