missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS51 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VPS51.
Brand: Abnova PAB25646.100ug
This item is not returnable.
View return policy
Specifications
| VPS51 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| VPS51 | |
| VPS51 | |
| A synthetic peptide corresponding to 18 amino acids at C-terminus of human VPS51. | |
| 100 μg | |
| Primary | |
| At least two alternatively spliced isoforms are known to exist; this antibody will detect both isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of VPS51. | |
| In PBS (0.05% sodium azide) | |
| PP5382/ANG2/ANG3/C11orf2/C11orf3/FFR | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 738 | |
| Human, Mouse, Rat | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction