Learn More
VPS13A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant VPS13A.
Brand: Abnova PAB21442.100uL
Description
The protein encoded by this gene may control steps in the cycling of proteins through the trans-Golgi network to endosomes, lysosomes and the plasma membrane. Mutations in this gene cause the autosomal recessive disorder, chorea-acanthocytosis. Alternative splicing of this gene results in multiple transcript variants. [provided by RefSeq
Sequence: RPPRFFNEDGVIRPYRLRDGTGNQMLQVMENGRFAKYKYFTHVMINKTDMLMITRRGVLFVTKGTFGQLTCEWQYSFDEFTKEPFIVHGRRLRIEAKERVKSVFSpecifications
| VPS13A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant VPS13A. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| CHAC/CHOREIN/FLJ42030/KIAA0986 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 23230 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| VPS13A | |
| VPS13A | |
| Recombinant protein corresponding to amino acids of human VPS13A. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only