Learn More
VIP Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of VIP.
Brand: Abnova PAB25075.100uL
Description
The protein encoded by this gene belongs to the glucagon family. It stimulates myocardial contractility, causes vasodilation, increases glycogenolysis, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq
Specifications
| VIP | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| VIP | |
| VIP | |
| A synthetic peptide corresponding to VIP. | |
| 100 μL | |
| Primary | |
| VIP polyclonal antibody detects endogenous levels of VIP protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of VIP. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| MGC13587/PHM27 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 7432 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only